The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276289.1 | NP_001263218.1 | 10 - 11 | 1201 | 85 |
NM_001276290.1 | NP_001263219.1 | 8 - 9 | 898 | 85 | |
NM_001278433.1 | NP_001265362.1 | 9 - 10 | 1071 | 85 | |
NM_002734.4 | NP_002725.1 | 9 - 10 | 1068 | 85 | |
NM_212471.2 | NP_997636.1 | 9 - 10 | 1261 | 85 | |
NM_212472.2 | NP_997637.1 | 9 - 10 | 1099 | 85 | |
XM_011524983.2 | XP_011523285.1 | 9 - 10 | 1164 | 85 | |
XM_011524984.2 | XP_011523286.1 | 10 - 11 | 1102 | 85 | |
XM_011524985.2 | XP_011523287.1 | 10 - 11 | 1396 | 85 | |
GenBank mRNA | A12295.1 | - | 9 - 10 | 994 | 85 |
AK097580.1 | - | 8 - 9 | 1471 | 85 | |
AK124586.1 | - | 9 - 10 | 987 | 85 | |
AK312307.1 | - | 9 - 10 | 997 | 85 | |
AW732104.1 | - | 2 - 3 | 85 | ||
BC036285.1 | - | 9 - 10 | 1020 | 85 | |
BC093042.1 | - | 9 - 10 | 1009 | 85 | |
CR749311.1 | - | 10 - 11 | 1130 | 85 | |
M18468.1 | - | 9 - 10 | 995 | 85 | |
M33336.1 | - | 9 - 10 | 979 | 85 | |
S54709.1 | - | 2 - 3 | 142 | 85 |