The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002734.4 | NP_002725.1 | 1 - 2 | 171 | 133 |
GenBank mRNA | BC036285.1 | - | 1 - 2 | 123 | 133 |
BM928493.1 | - | 1 - 2 | 169 | 133 | |
DA219203.1 | - | 1 - 2 | 171 | 133 | |
S54705.1 | - | 1 - 2 | 109 | 133 |