Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 12 - 13 2171 82
NM_144604.3 NP_653205.3 11 - 12 2099 82
XM_011522863.1 XP_011521165.1 10 - 11 1784 82
XM_011522865.1 XP_011521167.1 4 - 5 541 82
GenBank mRNA AK091849.1 - 6 - 7 926 82
AK225807.1 - 11 - 12 2069 82
AK300254.1 - 12 - 13 2149 82
AK302716.1 - 12 - 13 1545 82
BC001584.1 - 8 - 9 1195 82
BC050463.1 - 11 - 12 2046 82

View Full Product Details