Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 17 - 18 2837 148
NM_144604.3 NP_653205.3 16 - 17 2765 148
XM_011522863.1 XP_011521165.1 15 - 16 2450 148
XM_011522865.1 XP_011521167.1 9 - 10 1207 148
GenBank mRNA AK026317.1 - 2 - 3 292 148
AK091849.1 - 11 - 12 1592 148
AK093394.1 - 3 - 4 1017 148
AK225807.1 - 16 - 17 2735 148
AK300254.1 - 17 - 18 2815 148
BC001584.1 - 13 - 14 1861 148
BC050463.1 - 16 - 17 2712 148

View Full Product Details