Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 18 - 19 2934 82
NM_144604.3 NP_653205.3 17 - 18 2862 82
XM_011522863.1 XP_011521165.1 16 - 17 2547 82
XM_011522865.1 XP_011521167.1 10 - 11 1304 82
GenBank mRNA AK026317.1 - 3 - 4 389 82
AK091849.1 - 12 - 13 1689 82
AK093394.1 - 4 - 5 1114 82
AK225807.1 - 17 - 18 2832 82
AK300254.1 - 18 - 19 2912 82
BC001584.1 - 14 - 15 1958 82
BC050463.1 - 17 - 18 2809 82

View Full Product Details