Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 7 - 8 1365 80
NM_144604.3 NP_653205.3 6 - 7 1293 80
XM_017022934.1 XP_016878423.1 7 - 8 1365 80
GenBank mRNA AK091849.1 - 1 - 2 120 80
AK225807.1 - 6 - 7 1263 80
AK300254.1 - 7 - 8 1343 80
AK302716.1 - 6 - 7 920 80
BC001584.1 - 3 - 4 389 80
BC050463.1 - 6 - 7 1240 80
DB007291.1 - 3 - 4 379 80

View Full Product Details