Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 15 - 16 2540 133
NM_144604.3 NP_653205.3 14 - 15 2468 133
XM_011522863.1 XP_011521165.1 13 - 14 2153 133
XM_011522865.1 XP_011521167.1 7 - 8 910 133
GenBank mRNA AK091849.1 - 9 - 10 1295 133
AK093394.1 - 1 - 2 720 133
AK225807.1 - 14 - 15 2438 133
AK300254.1 - 15 - 16 2518 133
BC001584.1 - 11 - 12 1564 133
BC050463.1 - 14 - 15 2415 133

View Full Product Details