Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JOSD1
Entrez Gene ID
9929
Gene Name
Josephin domain containing 1
Gene Aliases
dJ508I15.2
Chromosome Location
Chr.22: 38685543 - 38701418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005261876.2 XP_005261933.1 1 - 2 143 60

View Full Product Details