Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAFR
Entrez Gene ID
5724
Gene Name
platelet activating factor receptor
Gene Aliases
PAFR
Chromosome Location
Chr.1: 28147166 - 28193936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000952.4 NP_000943.1 2 - 2 1393 88
NM_001164721.1 NP_001158193.1 3 - 3 1337 88
NM_001164722.2 NP_001158194.1 2 - 2 1255 88
NM_001164723.2 NP_001158195.1 3 - 3 1464 88
GenBank mRNA AK290240.1 - 2 - 2 1139 88
AK292053.1 - 3 - 3 1337 88
BC013816.2 - 1 - 1 1042 88
BC063000.1 - 2 - 2 1129 88
CD013879.1 - 3 - 3 1193 88
D10202.1 - 2 - 2 1114 88
M80436.1 - 2 - 2 1268 88

View Full Product Details