Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PYCRL
Entrez Gene ID
65263
Gene Name
pyrroline-5-carboxylate reductase-like
Gene Aliases
PYCR3
Chromosome Location
Chr.8: 143603913 - 143609614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023078.3 NP_075566.2 4 - 5 617 72
GenBank mRNA AK001500.2 - 4 - 5 554 72
AK023914.1 - 4 - 5 577 72
AK222685.1 - - 587 72
AK225404.1 - 4 - 5 566 72
AK294615.1 - 5 - 6 685 72
AK295444.1 - 4 - 5 558 72
AK301225.1 - 4 - 5 512 72
AL833857.1 - 3 - 4 461 72
BC007993.1 - 4 - 5 597 72

View Full Product Details