The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177562.1 | NP_001171033.1 | 5 - 6 | 767 | 132 |
NM_002716.4 | NP_002707.3 | 5 - 6 | 767 | 132 | |
NM_181699.2 | NP_859050.1 | 5 - 6 | 767 | 132 | |
NM_181700.1 | NP_859051.1 | 3 - 4 | 575 | 132 | |
XM_017017958.1 | XP_016873447.1 | 5 - 6 | 711 | 132 | |
XM_017017959.1 | XP_016873448.1 | 5 - 6 | 711 | 132 | |
XM_017017960.1 | XP_016873449.1 | 5 - 6 | 711 | 132 | |
XM_017017961.1 | XP_016873450.1 | 5 - 6 | 711 | 132 | |
GenBank mRNA | AF087438.1 | - | 5 - 6 | 712 | 132 |
AF163473.1 | - | 5 - 6 | 749 | 132 | |
AK292275.1 | - | 5 - 6 | 687 | 132 | |
AK293388.1 | - | 5 - 6 | 631 | 132 | |
AK294716.1 | - | 5 - 6 | 690 | 132 | |
AK301705.1 | - | 3 - 4 | 516 | 132 | |
BC027596.2 | - | 5 - 6 | 704 | 132 | |
M65254.1 | - | 5 - 6 | 605 | 132 |