Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECI2
Entrez Gene ID
10455
Gene Name
enoyl-CoA delta isomerase 2
Gene Aliases
ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3
Chromosome Location
Chr.6: 4115693 - 4135597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166010.1 NP_001159482.1 4 - 5 547 86
NM_006117.2 NP_006108.2 4 - 5 535 86
NM_206836.2 NP_996667.2 4 - 5 545 86
XM_006714957.1 XP_006715020.1 4 - 5 538 86
XM_017010170.1 XP_016865659.1 4 - 5 529 86
GenBank mRNA AB209917.1 - 3 - 4 529 86
AF069301.1 - 4 - 5 495 86
AF153612.1 - 4 - 5 505 86
AF244138.1 - 4 - 5 500 86
AF257175.1 - 4 - 5 497 86
AK057175.1 - 4 - 5 620 86
AK075108.1 - 4 - 5 513 86
AK222617.1 - 4 - 5 521 86
AK223003.1 - 4 - 5 523 86
AK297049.1 - 4 - 5 513 86
AL136642.1 - 4 - 5 522 86
AY297537.1 - 3 - 4 450 86
BC002668.2 - 4 - 5 522 86
BC016781.1 - 4 - 5 514 86
BC017474.1 - 4 - 5 499 86
BC033841.1 - 4 - 5 505 86
BC034702.1 - 4 - 5 547 86
CR456768.1 - 3 - 4 418 86

View Full Product Details