Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DVL3
Entrez Gene ID
1857
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
DRS3
Chromosome Location
Chr.3: 184155311 - 184173614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004423.3 NP_004414.3 10 - 11 1192 59
XM_005247172.2 XP_005247229.1 10 - 11 1377 59
XM_011512513.2 XP_011510815.1 7 - 8 980 59
GenBank mRNA AF006013.1 - 10 - 11 1063 59
AK093469.1 - 10 - 11 1132 59
AL110278.1 - 6 - 7 834 59
BC018289.1 - 8 - 9 728 59
BC032459.1 - 10 - 11 1120 59
D86963.1 - 10 - 11 1192 59
U48252.1 - 3 - 4 297 59
U49262.1 - 10 - 11 1173 59
U75651.1 - 10 - 11 1445 59

View Full Product Details