The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000158.3 | NP_000149.3 | 3 - 4 | 699 | 113 |
GenBank mRNA | AB209731.1 | - | 3 - 4 | 589 | 113 |
AK125918.1 | - | 3 - 4 | 1062 | 113 | |
AK300620.1 | - | 3 - 4 | 346 | 113 | |
AK312245.1 | - | 3 - 4 | 548 | 113 | |
BC012098.1 | - | 3 - 4 | 628 | 113 | |
L07956.1 | - | 3 - 4 | 506 | 113 |