Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKH
Entrez Gene ID
160851
Gene Name
diacylglycerol kinase eta
Gene Aliases
DGKeta
Chromosome Location
Chr.13: 42040036 - 42256584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204504.2 NP_001191433.1 3 - 4 407 71
NM_152910.5 NP_690874.2 2 - 3 370 71
NM_178009.4 NP_821077.1 2 - 3 370 71
GenBank mRNA AB078967.1 - 2 - 3 325 71
AB078968.1 - 2 - 3 325 71
AK289546.1 - 3 - 4 409 71
DR002010.1 - 2 - 3 378 71

View Full Product Details