The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323011.1 | NP_001309940.1 | 3 - 4 | 291 | 92 |
NM_001323012.1 | NP_001309941.1 | 3 - 4 | 298 | 92 | |
NM_006556.3 | NP_006547.1 | 3 - 4 | 641 | 92 | |
GenBank mRNA | AK312803.1 | - | 3 - 4 | 600 | 92 |
BC006089.1 | - | 3 - 4 | 624 | 92 | |
BC007694.2 | - | 3 - 4 | 554 | 92 | |
BQ061044.1 | - | 3 - 4 | 641 | 92 | |
BT019976.1 | - | 3 - 4 | 318 | 92 | |
CR541998.1 | - | 3 - 4 | 318 | 92 | |
L77213.1 | - | 3 - 4 | 353 | 92 |