The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321298.1 | NP_001308227.1 | 4 - 5 | 695 | 71 |
NM_173637.3 | NP_775908.2 | 3 - 4 | 597 | 71 | |
NR_135612.1 | 3 - 4 | 597 | 71 | ||
XM_011527926.1 | XP_011526228.1 | 3 - 4 | 391 | 71 | |
GenBank mRNA | AK097761.1 | - | 4 - 5 | 679 | 71 |
BC031671.1 | - | 3 - 4 | 597 | 71 |