The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024605.3 | NP_078881.3 | 13 - 14 | 1233 | 65 |
XM_005263215.3 | XP_005263272.1 | 13 - 14 | 1469 | 65 | |
XM_017008602.1 | XP_016864091.1 | 14 - 15 | 1505 | 65 | |
GenBank mRNA | AB050785.1 | - | 13 - 14 | 1233 | 65 |
AK123034.1 | - | 4 - 5 | 265 | 65 | |
AK304458.1 | - | 5 - 6 | 416 | 65 | |
BC011920.2 | - | 9 - 10 | 763 | 65 | |
BC109029.2 | - | 13 - 14 | 1223 | 65 | |
BC109030.1 | - | 13 - 14 | 1222 | 65 | |
BC126899.1 | - | 13 - 14 | 1229 | 65 | |
BC128055.1 | - | 13 - 14 | 1233 | 65 | |
BX647727.1 | - | 7 - 8 | 6363 | 65 |