The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024605.3 | NP_078881.3 | 20 - 21 | 2032 | 88 |
XM_017008602.1 | XP_016864091.1 | 21 - 22 | 2304 | 88 | |
GenBank mRNA | AB050785.1 | - | 20 - 21 | 2032 | 88 |
AK024549.1 | - | 2 - 3 | 332 | 88 | |
AK304458.1 | - | 12 - 13 | 1215 | 88 | |
AL834360.1 | - | 6 - 7 | 673 | 88 | |
BC011920.2 | - | 16 - 17 | 1562 | 88 | |
BC047914.1 | - | 6 - 7 | 1107 | 88 | |
BC106018.1 | - | 3 - 4 | 371 | 88 | |
BC109029.2 | - | 20 - 21 | 2022 | 88 | |
BC109030.1 | - | 20 - 21 | 2021 | 88 | |
BC126899.1 | - | 20 - 21 | 2028 | 88 | |
BC128055.1 | - | 20 - 21 | 2032 | 88 | |
BX647727.1 | - | 14 - 15 | 7162 | 88 | |
CR457328.1 | - | 2 - 3 | 331 | 88 |