Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP10
Entrez Gene ID
79658
Gene Name
Rho GTPase activating protein 10
Gene Aliases
GRAF2, PS-GAP, PSGAP
Chromosome Location
Chr.4: 147732063 - 148072781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024605.3 NP_078881.3 9 - 10 942 89
XM_005263215.3 XP_005263272.1 9 - 10 1178 89
XM_017008602.1 XP_016864091.1 10 - 11 1214 89
GenBank mRNA AB050785.1 - 9 - 10 942 89
AK304458.1 - 1 - 2 125 89
BC011920.2 - 5 - 6 472 89
BC109029.2 - 9 - 10 932 89
BC109030.1 - 9 - 10 931 89
BC126899.1 - 9 - 10 938 89
BC128055.1 - 9 - 10 942 89
BX647727.1 - 3 - 4 6072 89

View Full Product Details