Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP13
Entrez Gene ID
9319
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
16E1BP
Chromosome Location
Chr.5: 892854 - 919348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514163.2 XP_011512465.1 13 - 14 1467 77

View Full Product Details