Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP13
Entrez Gene ID
9319
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
16E1BP
Chromosome Location
Chr.5: 892854 - 919348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166260.1 NP_001159732.1 2 - 3 407 135
NM_004237.3 NP_004228.1 2 - 3 407 135
XM_011514163.2 XP_011512465.1 2 - 3 377 135
GenBank mRNA AK310376.1 - 2 - 3 377 135
BC000404.2 - 2 - 3 372 135
BC019294.1 - 2 - 3 372 135
BG722761.1 - 2 - 3 409 138
CR456744.1 - 2 - 3 262 135
L40384.1 - 2 - 3 268 135
U96131.1 - 2 - 3 307 135

View Full Product Details