Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP13
Entrez Gene ID
9319
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
16E1BP
Chromosome Location
Chr.5: 892854 - 919348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166260.1 NP_001159732.1 4 - 5 596 83
NM_004237.3 NP_004228.1 4 - 5 596 83
XM_011514163.2 XP_011512465.1 4 - 5 566 83
GenBank mRNA AK310376.1 - 4 - 5 566 83
BC000404.2 - 4 - 5 561 83
BC019294.1 - 4 - 5 561 83
CR456744.1 - 4 - 5 451 83
L40384.1 - 4 - 5 457 83
U96131.1 - 4 - 5 496 83

View Full Product Details