The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166260.1 | NP_001159732.1 | 8 - 9 | 905 | 53 |
NM_004237.3 | NP_004228.1 | 8 - 9 | 905 | 53 | |
XM_011514163.2 | XP_011512465.1 | 8 - 9 | 875 | 53 | |
GenBank mRNA | AK310376.1 | - | 8 - 9 | 875 | 53 |
BC000404.2 | - | 8 - 9 | 870 | 53 | |
BC019294.1 | - | 8 - 9 | 870 | 53 | |
CR456744.1 | - | 8 - 9 | 760 | 53 | |
U96131.1 | - | 8 - 9 | 805 | 53 |