The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207017.1 | NP_001193946.1 | 15 - 16 | 2340 | 92 |
NM_001207018.1 | NP_001193947.1 | 13 - 14 | 2269 | 92 | |
NM_006932.4 | NP_008863.3 | 14 - 15 | 2265 | 92 | |
NM_134269.2 | NP_599031.1 | 14 - 15 | 2265 | 92 | |
NM_134270.2 | NP_599032.2 | 14 - 15 | 2265 | 92 | |
XM_006724298.1 | XP_006724361.1 | 15 - 16 | 2336 | 92 | |
XM_006724302.1 | XP_006724365.1 | 15 - 16 | 2395 | 92 | |
XM_011530333.2 | XP_011528635.1 | 16 - 17 | 2441 | 92 | |
XM_011530341.1 | XP_011528643.1 | 8 - 9 | 1273 | 92 | |
XM_017028913.1 | XP_016884402.1 | 16 - 17 | 2442 | 92 | |
XM_017028914.1 | XP_016884403.1 | 15 - 16 | 2348 | 92 | |
XM_017028915.1 | XP_016884404.1 | 15 - 16 | 2348 | 92 | |
XM_017028916.1 | XP_016884405.1 | 16 - 17 | 2442 | 92 | |
XM_017028917.1 | XP_016884406.1 | 16 - 17 | 2442 | 92 | |
XM_017028918.1 | XP_016884407.1 | 16 - 17 | 2442 | 92 | |
GenBank mRNA | AB209643.1 | - | 9 - 10 | 3039 | 92 |
AF064238.3 | - | 14 - 15 | 2242 | 92 | |
AJ010306.2 | - | 14 - 15 | 2242 | 92 | |
AK095930.1 | - | 5 - 6 | 1678 | 92 | |
AK096953.1 | - | 14 - 15 | 2300 | 92 | |
AK128627.1 | - | 14 - 15 | 3903 | 92 | |
AK128719.1 | - | 13 - 14 | 3234 | 92 | |
AK225648.1 | - | 5 - 6 | 706 | 92 | |
AK293681.1 | - | 5 - 6 | 639 | 92 | |
AK295418.1 | - | 13 - 14 | 2269 | 92 | |
AK300064.1 | - | 5 - 6 | 786 | 92 | |
AK301289.1 | - | 4 - 5 | 541 | 92 | |
AK304084.1 | - | 15 - 16 | 2340 | 92 | |
AK307627.1 | - | 5 - 6 | 714 | 92 | |
AY061971.1 | - | 14 - 15 | 2243 | 92 | |
AY061972.1 | - | 14 - 15 | 2243 | 92 | |
BC034237.2 | - | 14 - 15 | 2233 | 92 | |
Y13492.2 | - | 14 - 15 | 2243 | 92 | |
Z49989.2 | - | 5 - 6 | 699 | 92 |