Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMTN
Entrez Gene ID
6525
Gene Name
smoothelin
Gene Aliases
-
Chromosome Location
Chr.22: 31081296 - 31104624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207017.1 NP_001193946.1 17 - 18 2475 107
NM_001207018.1 NP_001193947.1 15 - 16 2404 107
NM_006932.4 NP_008863.3 16 - 17 2400 107
NM_134269.2 NP_599031.1 16 - 17 2400 107
NM_134270.2 NP_599032.2 16 - 17 2400 107
XM_006724298.1 XP_006724361.1 17 - 18 2471 107
XM_006724302.1 XP_006724365.1 17 - 18 2530 107
XM_011530333.2 XP_011528635.1 18 - 19 2576 107
XM_011530336.2 XP_011528638.1 16 - 17 2349 107
XM_011530337.2 XP_011528639.1 16 - 17 2349 107
XM_011530338.2 XP_011528640.1 15 - 16 2255 107
XM_011530341.1 XP_011528643.1 10 - 11 1408 107
XM_017028913.1 XP_016884402.1 18 - 19 2577 107
XM_017028914.1 XP_016884403.1 17 - 18 2483 107
XM_017028915.1 XP_016884404.1 17 - 18 2483 107
XM_017028916.1 XP_016884405.1 18 - 19 2577 107
XM_017028917.1 XP_016884406.1 18 - 19 2577 107
XM_017028918.1 XP_016884407.1 17 - 18 2785 107
XM_017028919.1 XP_016884408.1 15 - 16 2255 107
GenBank mRNA AB209643.1 - 10 - 11 3382 107
AF064238.3 - 16 - 17 2377 107
AJ010306.2 - 16 - 17 2377 107
AK092684.1 - 4 - 5 818 107
AK095916.1 - 5 - 6 591 107
AK095930.1 - 6 - 7 2021 107
AK096953.1 - 15 - 16 2643 107
AK128627.1 - 16 - 17 4038 107
AK128719.1 - 14 - 15 3577 107
AK293681.1 - 6 - 7 982 107
AK295418.1 - 15 - 16 2404 107
AK301289.1 - 6 - 7 676 107
AK307627.1 - 6 - 7 1057 107
AY061971.1 - 16 - 17 2378 107
AY061972.1 - 16 - 17 2378 107
BC034237.2 - 16 - 17 2368 107
Y13492.2 - 16 - 17 2378 107
Z49989.2 - 7 - 8 834 107

View Full Product Details