The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173825.3 | NP_776186.2 | 6 - 7 | 640 | 80 |
GenBank mRNA | AK025772.1 | - | 3 - 4 | 336 | 80 |
AK293085.1 | - | 6 - 7 | 624 | 80 | |
BC020832.1 | - | 6 - 7 | 619 | 80 | |
BX647559.1 | - | 6 - 7 | 637 | 80 | |
BX647593.1 | - | 6 - 7 | 640 | 80 | |
BX647664.1 | - | 6 - 7 | 624 | 80 |