Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX2
Entrez Gene ID
55852
Gene Name
testis expressed 2
Gene Aliases
HT008, TMEM96
Chromosome Location
Chr.17: 64147433 - 64263323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288732.1 NP_001275661.1 10 - 11 3319 96
NM_001288733.1 NP_001275662.1 10 - 11 3311 96
NM_018469.4 NP_060939.3 10 - 11 3340 96
XM_011524998.1 XP_011523300.1 10 - 11 3329 96
XM_011524999.1 XP_011523301.1 10 - 11 3253 96
XM_011525000.1 XP_011523302.1 10 - 11 3304 96
XM_017024846.1 XP_016880335.1 10 - 11 3283 96
GenBank mRNA AB051525.2 - 9 - 10 3138 96
AF163260.1 - 8 - 9 1356 96
AK000369.1 - 5 - 6 645 96
AK000758.1 - 9 - 10 1555 96
AK091156.1 - 9 - 10 2308 96
AK314316.1 - 5 - 6 590 96
AL832371.1 - 10 - 11 3219 96
AL834251.2 - 10 - 11 3230 96
BC008896.2 - 9 - 10 2495 96
BC033661.1 - 7 - 8 1022 96
BC036672.1 - 10 - 11 3290 96
BC040521.1 - 10 - 11 3244 96
BC040522.1 - 10 - 11 3292 96
BC108279.1 - 8 - 9 1377 96
BX648260.1 - 2 - 3 1601 96
CR627433.1 - 10 - 11 3312 96

View Full Product Details