Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX2
Entrez Gene ID
55852
Gene Name
testis expressed 2
Gene Aliases
HT008, TMEM96
Chromosome Location
Chr.17: 64147433 - 64263323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288732.1 NP_001275661.1 5 - 6 2604 120
NM_001288733.1 NP_001275662.1 5 - 6 2596 120
NM_018469.4 NP_060939.3 5 - 6 2625 120
XM_011524998.1 XP_011523300.1 5 - 6 2614 120
XM_011524999.1 XP_011523301.1 5 - 6 2538 120
XM_011525000.1 XP_011523302.1 5 - 6 2595 120
XM_017024846.1 XP_016880335.1 5 - 6 2574 120
GenBank mRNA AB051525.2 - 4 - 5 2423 120
AF163260.1 - 3 - 4 641 120
AF220182.1 - 1 - 2 158 120
AK000758.1 - 4 - 5 840 120
AK091156.1 - 4 - 5 1593 120
AK091214.1 - 5 - 6 2521 120
AL832371.1 - 5 - 6 2504 120
AL834251.2 - 5 - 6 2515 120
BC008896.2 - 4 - 5 1781 120
BC033661.1 - 2 - 3 307 120
BC036672.1 - 5 - 6 2575 120
BC040521.1 - 5 - 6 2529 120
BC040522.1 - 5 - 6 2577 120
BC108279.1 - 3 - 4 662 120
CR627433.1 - 5 - 6 2597 120

View Full Product Details