The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135247.1 | NP_001128719.1 | 5 - 6 | 352 | 64 |
NM_001135248.1 | NP_001128720.1 | 5 - 6 | 371 | 64 | |
NM_002403.3 | NP_002394.1 | 5 - 6 | 374 | 64 | |
NM_017459.2 | NP_059453.1 | 5 - 6 | 355 | 64 | |
XM_011541476.2 | XP_011539778.1 | 5 - 6 | 981 | 64 | |
GenBank mRNA | AK222751.1 | - | 5 - 6 | 333 | 64 |
AK301655.1 | - | 4 - 5 | 449 | 64 | |
BC015039.1 | - | 5 - 6 | 374 | 64 | |
CR407678.1 | - | 4 - 5 | 241 | 64 | |
DN996554.1 | - | 5 - 6 | 324 | 64 | |
U19718.1 | - | 4 - 5 | 282 | 64 |