Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
Chromosome Location
Chr.1: 16974502 - 16981649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135247.1 NP_001128719.1 7 - 8 487 82
NM_001135248.1 NP_001128720.1 7 - 8 506 82
NM_002403.3 NP_002394.1 7 - 8 509 82
NM_017459.2 NP_059453.1 7 - 8 490 82
XM_011541476.2 XP_011539778.1 7 - 8 1116 82
GenBank mRNA AK222751.1 - 7 - 8 468 82
BC015039.1 - 7 - 8 509 82
CR407678.1 - 6 - 7 376 82
DN996554.1 - 7 - 8 459 82
U19718.1 - 6 - 7 417 82

View Full Product Details