Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
Chromosome Location
Chr.1: 16974502 - 16981649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135247.1 NP_001128719.1 8 - 9 563 105
NM_001135248.1 NP_001128720.1 8 - 9 582 105
NM_002403.3 NP_002394.1 8 - 9 585 105
NM_017459.2 NP_059453.1 8 - 9 566 105
XM_011541476.2 XP_011539778.1 8 - 9 1192 105
GenBank mRNA AK222751.1 - 8 - 9 544 105
BC015039.1 - 8 - 9 585 105
CR407678.1 - 7 - 8 452 105
DN996554.1 - 8 - 9 535 105
U19718.1 - 7 - 8 493 105

View Full Product Details