Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GM2A
Entrez Gene ID
2760
Gene Name
GM2 ganglioside activator
Gene Aliases
GM2-AP, SAP-3
Chromosome Location
Chr.5: 151253052 - 151270394 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000405.4 NP_000396.2 3 - 4 597 111
GenBank mRNA AK225978.1 - 3 - 4 525 111
AK312494.1 - 3 - 4 492 111
BC009273.2 - 3 - 4 461 111
L01439.1 - 3 - 4 522 111
M76477.1 - 3 - 4 522 111
X16087.1 - 3 - 4 390 111
X61095.1 - 3 - 4 453 111
X62078.1 - 3 - 4 490 111

View Full Product Details