The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180991.4 | NP_851322.3 | 8 - 9 | 1759 | 89 |
XM_011543370.1 | XP_011541672.1 | 7 - 8 | 1437 | 89 | |
XM_011543372.1 | XP_011541674.1 | 10 - 11 | 1410 | 89 | |
GenBank mRNA | AF401643.1 | - | 8 - 9 | 1472 | 89 |
AK128854.1 | - | 8 - 9 | 1487 | 89 | |
AY273896.2 | - | 8 - 9 | 1759 | 89 | |
BX647349.1 | - | 8 - 9 | 1652 | 89 |