Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11orf80
Entrez Gene ID
79703
Gene Name
chromosome 11 open reading frame 80
Gene Aliases
TOP6BL, TOPOVIBL
Chromosome Location
Chr.11: 66744736 - 66843516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302084.1 NP_001289013.1 5 - 6 489 97
NM_024650.3 NP_078926.3 7 - 8 781 97
GenBank mRNA AK026184.1 - 6 - 7 582 97
AK225435.1 - 6 - 7 582 97
AK302025.1 - 5 - 6 441 97
BC028240.1 - 7 - 8 784 97

View Full Product Details