Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASQ1
Entrez Gene ID
844
Gene Name
calsequestrin 1
Gene Aliases
CASQ, PDIB1, VMCQA
Chromosome Location
Chr.1: 160190495 - 160201886 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001231.4 NP_001222.3 2 - 3 618 127
GenBank mRNA AK313250.1 - 2 - 3 521 127
BC022289.1 - 2 - 3 504 127
S73775.1 - 2 - 3 508 127

View Full Product Details