The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244644.1 | NP_001231573.1 | 2 - 3 | 75 | |
NM_014043.3 | NP_054762.2 | 3 - 4 | 75 | ||
XM_011533576.2 | XP_011531878.1 | 3 - 4 | 75 | ||
GenBank mRNA | AF151842.1 | - | 3 - 4 | 542 | 75 |
AK002165.1 | - | 3 - 4 | 556 | 75 | |
AK002180.1 | - | 3 - 4 | 561 | 75 | |
AK296072.1 | - | 2 - 3 | 483 | 75 | |
AK316585.1 | - | 3 - 4 | 547 | 75 | |
AL080122.1 | - | 3 - 4 | 556 | 75 | |
BC001553.1 | - | 3 - 4 | 399 | 75 | |
BI821848.1 | - | 3 - 4 | 75 | ||
BM828410.1 | - | 3 - 4 | 583 | 75 | |
CR533456.1 | - | 3 - 4 | 323 | 75 |