The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178044.1 | NP_001171515.1 | 11 - 12 | 1070 | 83 |
NM_001178045.1 | NP_001171516.1 | 12 - 13 | 1199 | 83 | |
NM_025257.2 | NP_079533.2 | 12 - 13 | 1196 | 83 | |
GenBank mRNA | AK027397.1 | - | 12 - 13 | 1147 | 83 |
AK222998.1 | - | 12 - 13 | 1147 | 83 | |
AK300550.1 | - | 12 - 13 | 1199 | 83 | |
AK301596.1 | - | 11 - 12 | 1021 | 83 | |
AK314673.1 | - | 12 - 13 | 1151 | 83 | |
AL833009.1 | - | 8 - 9 | 889 | 83 | |
BC014659.1 | - | 12 - 13 | 1135 | 83 | |
KU175229.1 | - | 12 - 13 | 1130 | 83 |