The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145415.1 | NP_001138887.1 | 18 - 19 | 3482 | 58 |
NM_012432.3 | NP_036564.3 | 18 - 19 | 3482 | 58 | |
XM_005245641.3 | XP_005245698.1 | 18 - 19 | 3485 | 58 | |
XM_017002953.1 | XP_016858442.1 | 18 - 19 | 3409 | 58 | |
GenBank mRNA | AK095075.1 | - | 2 - 3 | 976 | 58 |
BC028671.2 | - | 18 - 19 | 3409 | 58 | |
D31891.1 | - | 18 - 19 | 3378 | 58 |