Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGB2
Entrez Gene ID
3689
Gene Name
integrin subunit beta 2
Gene Aliases
CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7
Chromosome Location
Chr.21: 44885949 - 44928873 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000211.4 NP_000202.3 14 - 15 2309 66
NM_001127491.2 NP_001120963.2 14 - 15 2299 66
NM_001303238.1 NP_001290167.1 14 - 15 2264 66
XM_006724001.1 XP_006724064.1 14 - 15 2326 66
XM_017028341.1 XP_016883830.1 14 - 15 2321 66
GenBank mRNA AB208909.1 - 13 - 14 4132 66
AK095992.1 - 14 - 15 2264 66
AK097216.1 - 5 - 6 1476 66
AK128344.1 - 14 - 15 2134 66
AK222505.1 - 14 - 15 2166 66
AK303488.1 - 14 - 15 2177 66
BC005861.2 - 13 - 14 2120 66
BC021077.1 - 13 - 14 2043 66
CA447686.1 - 2 - 3 66
L78790.1 - 13 - 14 1956 66
M15395.1 - 14 - 15 2148 66
Y00057.1 - 13 - 14 2052 66

View Full Product Details