Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
Chromosome Location
Chr.2: 227472169 - 227561222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135187.1 NP_001128659.1 3 - 4 632 111
NM_001135188.1 NP_001128660.1 3 - 4 632 111
NM_001135189.1 NP_001128661.1 3 - 4 632 111
NM_004504.4 NP_004495.2 3 - 4 632 111
XM_005246516.3 XP_005246573.1 3 - 4 472 111
XM_006712478.3 XP_006712541.1 3 - 4 472 111
XM_006712479.3 XP_006712542.1 3 - 4 472 111
XM_006712480.3 XP_006712543.1 3 - 4 635 111
GenBank mRNA AI742626.1 - - 480 111
AK097451.1 - 4 - 5 527 111
AK295030.1 - 2 - 3 182 111
BC030592.2 - 3 - 4 466 111
BC096272.3 - 3 - 4 432 111
BC096273.3 - 3 - 4 432 111
BC096274.1 - 3 - 4 432 111
BC096275.3 - 3 - 4 432 111
KU178123.1 - 3 - 4 382 111
KU178124.1 - 3 - 4 382 111
KU178125.1 - 3 - 4 382 111
KU178126.1 - 3 - 4 382 111
X89478.1 - 3 - 4 448 111

View Full Product Details