The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024782.2 | NP_079058.1 | 2 - 3 | 322 | 67 |
GenBank mRNA | AJ972687.1 | - | 2 - 3 | 266 | 67 |
AK022672.1 | - | 2 - 3 | 258 | 67 | |
BC012732.2 | - | 2 - 3 | 243 | 67 | |
BC030986.1 | - | 2 - 3 | 275 | 67 | |
CR457291.1 | - | 1 - 2 | 176 | 67 |