Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAI
Entrez Gene ID
286205
Gene Name
suppressor of cancer cell invasion
Gene Aliases
C9orf126, NET40
Chromosome Location
Chr.9: 124942608 - 125143559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173690.4 NP_775961.2 3 - 4 285 103
GenBank mRNA AK098024.1 - 3 - 4 222 103
BC104030.2 - 3 - 4 213 103

View Full Product Details