The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145415.1 | NP_001138887.1 | 11 - 12 | 1618 | 79 |
NM_012432.3 | NP_036564.3 | 11 - 12 | 1618 | 79 | |
GenBank mRNA | BC028671.2 | - | 11 - 12 | 1545 | 79 |
D31891.1 | - | 11 - 12 | 1514 | 79 |