The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168647.1 | NP_001162118.1 | 10 - 11 | 1566 | 77 |
NM_001168648.1 | NP_001162119.1 | 10 - 11 | 1566 | 77 | |
NM_001168649.1 | NP_001162120.1 | 9 - 10 | 1419 | 77 | |
NM_014927.3 | NP_055742.2 | 10 - 11 | 1566 | 77 | |
XM_011545471.2 | XP_011543773.1 | 10 - 11 | 1649 | 77 | |
XM_011545472.2 | XP_011543774.1 | 9 - 10 | 1423 | 77 | |
XM_017029358.1 | XP_016884847.1 | 9 - 10 | 1423 | 77 | |
GenBank mRNA | AB020709.1 | - | 10 - 11 | 1542 | 77 |
AF418269.1 | - | 10 - 11 | 1122 | 77 | |
AF418270.1 | - | 10 - 11 | 1122 | 77 | |
AK294728.1 | - | 9 - 10 | 1381 | 77 | |
BC126121.1 | - | 10 - 11 | 1121 | 77 | |
BC136289.1 | - | 10 - 11 | 1121 | 77 | |
BC143839.1 | - | 10 - 11 | 1121 | 77 |