The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168648.1 | NP_001162119.1 | 11 - 12 | 1783 | 81 |
NM_001168649.1 | NP_001162120.1 | 10 - 11 | 1636 | 81 | |
NM_014927.3 | NP_055742.2 | 11 - 12 | 1783 | 81 | |
XM_011545472.2 | XP_011543774.1 | 10 - 11 | 1640 | 81 | |
GenBank mRNA | AB020709.1 | - | 11 - 12 | 1759 | 81 |
AF418269.1 | - | 11 - 12 | 1339 | 81 | |
AF418270.1 | - | 11 - 12 | 1339 | 81 | |
AK294728.1 | - | 10 - 11 | 1598 | 81 | |
BC126121.1 | - | 11 - 12 | 1338 | 81 | |
BC136289.1 | - | 11 - 12 | 1338 | 81 |