Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNKSR2
Entrez Gene ID
22866
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
CNK2, KSR2, MAGUIN
Chromosome Location
Chr.X: 21374335 - 21654695 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168647.1 NP_001162118.1 3 - 4 911 93
NM_001168648.1 NP_001162119.1 3 - 4 911 93
NM_001168649.1 NP_001162120.1 3 - 4 911 93
NM_014927.3 NP_055742.2 3 - 4 911 93
XM_011545471.2 XP_011543773.1 3 - 4 994 93
XM_011545472.2 XP_011543774.1 3 - 4 915 93
XM_017029358.1 XP_016884847.1 3 - 4 915 93
GenBank mRNA AB020709.1 - 3 - 4 887 93
AF418269.1 - 3 - 4 467 93
AF418270.1 - 3 - 4 467 93
AK294728.1 - 3 - 4 873 93
BC126121.1 - 3 - 4 466 93
BC136289.1 - 3 - 4 466 93
BC143839.1 - 3 - 4 466 93

View Full Product Details