Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC00597
- Entrez Gene ID
-
81698
- Gene Name
- long intergenic non-protein coding RNA 597
- Gene Aliases
-
C15orf5
- Chromosome Location
-
Chr.15: 77223893 - 77225404 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026813.1 |
|
1 - 1 |
957 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
pseudopodium enriched atypical kinase 1 |
PEAK1 |
SGK269 |
|
Sense |
Chr. - 77100829 - 77420104 on Build GRCh38 |
Downstream |
high mobility group 20A |
HMG20A |
HMGX1, HMGXB1 |
195.2kb
|
Anti-sense |
Chr. - 77420651 - 77519899 on Build GRCh38 |
View Full Product Details