Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
Chromosome Location
Chr.9: 104781002 - 104928246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005502.3 NP_005493.2 4 - 5 710 67
XM_005251773.2 XP_005251830.1 4 - 5 619 67
XM_005251776.2 XP_005251833.1 3 - 4 459 67
XM_011518339.2 XP_011516641.1 5 - 6 694 67
XM_011518340.2 XP_011516642.1 6 - 7 909 67
XM_011518341.2 XP_011516643.1 5 - 6 694 67
XM_011518344.2 XP_011516646.1 5 - 6 694 67
XM_017014378.1 XP_016869867.1 5 - 6 892 67
XM_017014379.1 XP_016869868.1 5 - 6 1300 67
XM_017014380.1 XP_016869869.1 5 - 6 626 67
XM_017014381.1 XP_016869870.1 5 - 6 692 67
XM_017014382.1 XP_016869871.1 3 - 4 1157 67
GenBank mRNA AB055982.1 - 3 - 4 306 67
AB445477.1 - 3 - 4 306 67
AF165281.1 - 2 - 3 246 67
AF285167.1 - 4 - 5 596 67
AJ012376.1 - 2 - 3 246 67
AK024328.1 - 4 - 5 619 67
BC146856.1 - 4 - 5 520 67

View Full Product Details