Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
Chromosome Location
Chr.9: 104781002 - 104928246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005502.3 NP_005493.2 5 - 6 822 80
XM_005251773.2 XP_005251830.1 5 - 6 731 80
XM_005251776.2 XP_005251833.1 4 - 5 571 80
XM_011518339.2 XP_011516641.1 6 - 7 806 80
XM_011518340.2 XP_011516642.1 7 - 8 1021 80
XM_011518341.2 XP_011516643.1 6 - 7 806 80
XM_011518342.2 XP_011516644.1 3 - 4 509 80
XM_011518344.2 XP_011516646.1 6 - 7 806 80
XM_017014378.1 XP_016869867.1 6 - 7 1004 80
XM_017014379.1 XP_016869868.1 6 - 7 1412 80
XM_017014380.1 XP_016869869.1 6 - 7 738 80
XM_017014381.1 XP_016869870.1 6 - 7 804 80
XM_017014382.1 XP_016869871.1 4 - 5 1269 80
GenBank mRNA AB055982.1 - 4 - 5 418 80
AB445477.1 - 4 - 5 418 80
AF165281.1 - 3 - 4 358 80
AF285167.1 - 5 - 6 708 80
AJ012376.1 - 3 - 4 358 80
AK024328.1 - 5 - 6 731 80
BC146856.1 - 5 - 6 632 80

View Full Product Details